missing translation for 'onlineSavingsMsg'
Learn More

Atovaquone 98.0+%, TCI America™

Catalog No. a25451g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
A25451G 1 g
A2545200MG 200 mg
2 options

Catalog No. A25451G

Supplier: TCI America A25451G

Only null left

Chemical Identifiers

CAS 95233-18-4
Molecular Formula C22H19ClO3
Molecular Weight (g/mol) 366.84
MDL Number MFCD00889188
InChI Key BSJMWHQBCZFXBR-UHFFFAOYSA-N
Synonym trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione
PubChem CID 74989
IUPAC Name 3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione
SMILES OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 223°C
Quantity 1 g
Formula Weight 366.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Atovaquone
RTECSNumber QJ5616500
TSCA No
Recommended Storage Freezer