missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Related Compound D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013165
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013165 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013165 Supplier Merck Emd Millipore Supplier No. PHR187130MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Atorvastatin Related Compound H is an impurity of Atorvastatin, an inhibitor of 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase, an enzyme that catalyzes the conversion of HMG-CoA to mevalonate in cholesterol biosynthesis pathway.

Specifications

CAS 148146-51-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C26H22 FNO4
Synonym 3-(4-Fluorobenzoyl)-2-(2-methyl-1-oxopropyl)-N,3-diphenyloxiranecarboxamide
Type Certified Reference Material
Quantity 30 mg
Formula Weight 431.46
Grade Certified Reference Material