missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015259
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015259 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015259 Supplier Merck Emd Millipore Supplier No. PHR187030MG
This material is an impurity of atorvastatin. Atorvastatin is a synthetic HMG-CoA reductase inhibitor which lowers plasma cholesterol levels by inhibiting endogenous cholesterol synthesis.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 693793-53-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C66H66 CaF4N4O10
Synonym (betaR,deltaR)-2,3-Bis(4-fluorophenyl)-beta,delta-dihydroxy-5-(1-methylethyl)-4-[(phenylamino)carbonyl]-1 H-pyrrole-1-heptanoic acid hemicalcium salt
Type Certified Reference Material
Quantity 30 mg
Formula Weight 1191.32
Grade Certified Reference Material