missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Lactone 95.0+%, TCI America™

Catalog No. p-7118224
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A311425MG 25 mg
A3114100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A311425MG Supplier TCI America Supplier No. A311425MG
Only null left

Chemical Identifiers

CAS 125995-03-1
Molecular Formula C33H33FN2O4
Molecular Weight (g/mol) 540.635
MDL Number MFCD00899262
InChI Key OUCSEDFVYPBLLF-KAYWLYCHSA-N
Synonym 5-(4-Fluorophenyl)-1-[2-[(2R,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-2-isopropyl-N,4-diphenyl-1H-pyrrole-3-carboxamide, Atorvastatin Related Compound H
PubChem CID 6483036
IUPAC Name 5-(4-fluorophenyl)-1-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-N,4-diphenyl-2-propan-2-ylpyrrole-3-carboxamide
SMILES CC(C)C1=C(C(=C(N1CCC2CC(CC(=O)O2)O)C3=CC=C(C=C3)F)C4=CC=CC=C4)C(=O)NC5=CC=CC=C5

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 25 mg
Formula Weight 540.64
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Atorvastatin Lactone