missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Atorvastatin calcium trihydrate

Chemical Identifiers

CAS 344423-98-9
Molecular Formula C66H74CaF2N4O13
Molecular Weight (g/mol) 1209.41
InChI Key SHZPNDRIDUBNMH-NIJVSVLQSA-L
IUPAC Name calcium bis((3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoate) trihydrate
SMILES O.O.O.[Ca++].CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1.CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC464810010
View Documents
Thermo Scientific™
464810010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC464810050
View Documents
Thermo Scientific™
464810050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

344423-98-9
1209.41
calcium bis((3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoate) trihydrate
C66H74CaF2N4O13
SHZPNDRIDUBNMH-NIJVSVLQSA-L
O.O.O.[Ca++].CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1.CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1

Specifications

344423-98-9
1 g
SHZPNDRIDUBNMH-NIJVSVLQSA-L
calcium bis((3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoate) trihydrate
604.69
C66H74CaF2N4O13
Solubility aq. soln.: insoluble. Other solubilities: soluble in dmso, ethanol
O.O.O.[Ca++].CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1.CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1
1209.41
Atorvastatin calcium trihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye irritation.

May damage fertility or the unborn child.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only