missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Atorvastatin calcium trihydrate

Catalog No. AC464810010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464810010 1 g
AC464810050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464810010 Supplier Thermo Scientific Chemicals Supplier No. 464810010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 344423-98-9 | C66H74CaF2N4O13 | 1209.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 344423-98-9
Molecular Formula C66H74CaF2N4O13
Molecular Weight (g/mol) 1209.41
InChI Key SHZPNDRIDUBNMH-NIJVSVLQSA-L
IUPAC Name calcium bis((3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoate) trihydrate
SMILES O.O.O.[Ca++].CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1.CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1

Specifications

Melting Point 156.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in dmso, ethanol
Formula Weight 604.69
Chemical Name or Material Atorvastatin calcium trihydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye irritation.

May damage fertility or the unborn child.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only