missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Calcium Salt Trihydrate 98.0+%, TCI America™

Catalog No. A24761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24761G 1 g
A24765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24761G Supplier TCI America Supplier No. A24761G

Chemical Identifiers

CAS 344423-98-9
Molecular Formula C66H74CaF2N4O13
Molecular Weight (g/mol) 1209.408
MDL Number MFCD09752074
InChI Key SHZPNDRIDUBNMH-NIJVSVLQSA-L
PubChem CID 656846
ChEBI CHEBI:2911
IUPAC Name calcium;(3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate;trihydrate
SMILES CC(C)C1=C(C(=C(N1CCC(CC(CC(=O)[O-])O)O)C2=CC=C(C=C2)F)C3=CC=CC=C3)C(=O)NC4=CC=CC=C4.CC(C)C1=C(C(=C(N1CCC(CC(CC(=O)[O-])O)O)C2=CC=C(C=C2)F)C3=CC=CC=C3)C(=O)NC4=CC=CC=C4.O.O.O.[Ca+2]

Specifications

Color White
Quantity 1 g
Formula Weight 1155.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Atorvastatin Calcium Salt Trihydrate