missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Acetonide tert-Butyl Ester 98.0+%, TCI America™

Catalog No. p-7123106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36901G 1 g
B36905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36901G Supplier TCI America Supplier No. B36901G
Only null left

Chemical Identifiers

CAS 125971-95-1
Molecular Formula C40H47FN2O5
Molecular Weight (g/mol) 654.823
MDL Number MFCD04039904
InChI Key NPPZOMYSGNZDKY-ROJLCIKYSA-N
Synonym Atorvastatin Acetonide tert-Butyl Ester
PubChem CID 10168503
IUPAC Name tert-butyl 2-[(4R,6R)-6-[2-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMILES CC(C)C1=C(C(=C(N1CCC2CC(OC(O2)(C)C)CC(=O)OC(C)(C)C)C3=CC=C(C=C3)F)C4=CC=CC=C4)C(=O)NC5=CC=CC=C5

Specifications

Melting Point 146°C
Color White
Quantity 1 g
Formula Weight 654.82
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl (4R,6R)-2-[6-[2-[2-(4-Fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]e