missing translation for 'onlineSavingsMsg'
Learn More

Atomoxetine Hydrochloride 98.0+%, TCI America™

Catalog No. A2357100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2357100MG 100 mg
A23571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2357100MG Supplier TCI America Supplier No. A2357100MG

Chemical Identifiers

CAS 82248-59-7
Molecular Formula C17H22ClNO
Molecular Weight (g/mol) 291.819
MDL Number MFCD06410992
InChI Key LUCXVPAZUDVVBT-UNTBIKODSA-N
Synonym atomoxetine hcl, atomoxetine hydrochloride, strattera, tomoxetine hydrochloride, r-tomoxetine hydrochloride, tomoxetine, r-n-methyl-3-phenyl-3-o-tolyloxy propan-1-amine hydrochloride, r---tomoxetine hydrochloride, unii-57wvb6i2w0
PubChem CID 54840
ChEBI CHEBI:331697
IUPAC Name (3R)-N-methyl-3-(2-methylphenoxy)-3-phenylpropan-1-amine;hydrochloride
SMILES CC1=CC=CC=C1OC(CCNC)C2=CC=CC=C2.Cl

Specifications

Melting Point 169°C
Color White
Quantity 100 mg
Formula Weight 291.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Atomoxetine Hydrochloride
TSCA No
Recommended Storage Freezer