missing translation for 'onlineSavingsMsg'
Learn More

Atomoxetine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015620
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015620 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015620 Supplier Merck Emd Millipore Supplier No. PHR1679500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Atomoxetine hydrochloride is a nonstimulant used in the treatment of Attention-Deficit/Hyperactivity Disorder (ADHD) in children and adults.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 82248-59-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H21 NO · HCl
MDL Number MFCD06410992
Synonym (R)-Tomoxetine hydrochloride; (R)-N-Methyl-gamma-(2-methyl­phenoxy)benzenepropanamine hydrochloride; Atomoxetine hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 291.82
Grade Certified Reference Material