missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ Triple-Dipped PVC Gloves with Aramid Fiber Lining
Gloves provide protection against a wide range of hazards including cuts, abrasions, chemicals, and oils. Rough textured surface increases durability and enhances grip capabilities.
$181.44 - $196.77
Specifications
| ANSI Abrasion Rating | ANSI 6 |
|---|---|
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Coating Material | PVC |
| Color | Blue |
| Cut Resistance Rating | ANSI A2 |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-014-383
|
SHOWA™
KV660M08 |
Medium |
Dozen of 12 Each for $181.44
Case of 6 Dozen for $1,088.64
|
|
|||||
|
19-014-384
|
SHOWA™
KV660L09 |
Large |
Dozen of 12 Each for $196.77
Case of 6 Dozen for $1,180.62
|
|
|||||
|
19-014-385
|
SHOWA™
KV660XL10 |
X-Large |
Dozen of 12 Each for $181.44
Case of 6 Dozen for $1,088.64
|
|
|||||
|
19-014-386
|
SHOWA™
KV660XXL11 |
2X-Large |
Dozen of 12 Each for $181.44
Case of 6 Dozen for $1,088.64
|
|
|||||
Description
- Triple dipped PVC coating protects against oils and chemicals
- Kevlar™ aramid fiber liner provides cut resistance
- Rough textured surface enhances grip capabilities
- Hazard barrier is maintained even after multiple washes
- ANSI/ISEA 105-16 Level 4 Puncture Resistance
Specifications
| ANSI 6 | |
| PVC | |
| ANSI A2 | |
| Specific | |
| 4X42B | |
| Chemical Spray, Coating Preparation, Drainage, HVAC, Plumbing, and Painting | |
| Aramid Fiber | |
| 12 in. |
| EN 420:2003 + A1:2009 | |
| Blue | |
| Fully Textured | |
| Dip-coated Gloves | |
| JKL | |
| Open | |
| KV660 | |
| 30.5 cm |