missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ Triple-Dipped PVC Gloves with Aramid Fiber Lining
Gloves provide protection against a wide range of hazards including cuts, abrasions, chemicals, and oils. Rough textured surface increases durability and enhances grip capabilities.
Supplier: SHOWA™ KV660XXL11
Description
- Triple dipped PVC coating protects against oils and chemicals
- Kevlar™ aramid fiber liner provides cut resistance
- Rough textured surface enhances grip capabilities
- Hazard barrier is maintained even after multiple washes
- ANSI/ISEA 105-16 Level 4 Puncture Resistance
Specifications
| ANSI 6 | |
| PVC | |
| ANSI A2 | |
| Specific | |
| 2X-Large | |
| JKL | |
| Open | |
| KV660 | |
| 30.5 cm |
| EN 420:2003 + A1:2009 | |
| Blue | |
| Fully Textured | |
| Dip-coated Gloves | |
| 4X42B | |
| Chemical Spray, Coating Preparation, Drainage, HVAC, Plumbing, and Painting | |
| Aramid Fiber | |
| 12 in. |