missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATGL Monoclonal Antibody (K.751.7)

Description
It is not recommended to aliquot this antibody.
Lipolytic enzymes are required for mobilization of fatty acids from triglyceride stores in adipose tissue. Energy homeostasis is affected by dysfunctional lipolysis and may contribute to the pathogenesis of obesity and insulin resistance. Until recently, hormone-sensitive lipase (HSL) was the only enzyme known to hydrolyze triglycerides in mammalian adipose tissue. It is now thought that a second enzyme, adipose triglyceride lipase (ATGL), catalyzes the initial step in triglyceride hydrolysis. ATGL is highly expressed in adipose tissue of mice and humans. It exhibits high substrate specificity for triacylglycerol and is associated with lipid droplets. Inhibition of ATGL markedly decreases total adipose acyl-hydrolase activity. Thus, ATGL and HSL coordinately catabolize stored triglycerides in adipose tissue of mammals.
Specifications
Specifications
| Antigen | ATGL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | K.751.7 |
| Concentration | 23 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 |
| Gene | PNPLA2 |
| Gene Accession No. | P0C548, Q8BJ56 |
| Gene Alias | 0610039C21Rik; 1110001C14Rik; adipose triglyceride lipase; ATGL; ATGL DESNUTRIN; calcium-independent phospholipase A2; Desnutrin; DKFZp667M109; FP17548; IPLA2 zeta; iPLA2zeta; IPLA2-zeta; mutant patatin-like phospholipase domain containing 2; patatin like phospholipase domain containing 2; patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing protein 2; patatin-like phospholipase domain-containing protein 2; PEDF R; PEDF-R; pigment epithelium-derived factor; plpl; plpl2; Pnpla2; RGD1309044; transport-secretion protein; Transport-secretion protein 2; transport-secretion protein 2.2; triglyceride hydrolase; TTS 2.2; TTS2; TTS2.2; TTS-2.2 |
| Show More |