missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATG7 Recombinant Rabbit Monoclonal Antibody (23GB1730)

Catalog No. MA552681
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552681 20 μL
MA552680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552681 Supplier Invitrogen™ Supplier No. MA552681
Only null left

Rabbit Recombinant Monoclonal Antibody

ATG7 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vacuolar membranes. The protein shows homology to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.

Specifications

Antigen ATG7
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1730
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATG7
Gene Accession No. O95352, Q641Y5, Q9D906
Gene Alias 1810013K23Rik; Agp7; APG7; APG7L; APG7-LIKE; AT5G45900; ATAPG7; ATATG7; ATG12-activating enzyme E1 ATG7; Atg7; ATG7 autophagy related 7 homolog; Atg7l; AUTOPHAGY 7; autophagy related 7; autophagy related 7 homolog; autophagy-related 7; autophagy-related protein 7; DKFZp434N0735; Gm21553; GSA7; hAGP7; K15I22.10; K15I22_10; PEROXISOME UNUSUAL POSITIONING 4; PEUP4; ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Gene Symbols ATG7
Host Species Rabbit
Immunogen A synthesized peptide derived from human Atg7 (Apg7) (2-65AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10533, 312647, 74244
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less