missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATG4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106650 Supplier Invitrogen™ Supplier No. PA5106650
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATG4D.

Atg4D (autophagy-related gene 4D), also known as APG4D or AUTL4, is a 474 amino acid protein that localizes to the cytoplasm and belongs to the C-54 family of cysteine proteases. Expressed predominately in skeletal muscle, but also present in testis, Atg4D functions as a cysteine protease that is required for autophagy and functions to specifically cleave the C-terminal region of target proteins, thereby allowing the target proteins to bind to autophagosomes. The enzymatic activity of Atg4D is inhibited by Nethylmaleimide, a thiol reactive compound that is capable of modifying cystine residues in proteins and peptides.

Specifications

Antigen ATG4D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Atg4d
Gene Accession No. Q86TL0, Q8BGV9
Gene Alias 9830134P12Rik; APG4 (ATG4) autophagy-related homolog D; APG4 autophagy 4 homolog D; APG4D; APG4-D; ATG4 autophagy related 4 homolog D; Atg4d; Atg4dl; Autl4; AUT-like 4 cysteine endopeptidase; AUT-like 4, cysteine endopeptidase; Autophagin-4; autophagy related 4D cysteine peptidase; autophagy related 4D, cysteine peptidase; autophagy-related 4D; autophagy-related cysteine endopeptidase 4; Autophagy-related protein 4 homolog D; cysteine protease ATG4D; Cysteine protease ATG4D, mitochondrial; cysteine protease involved in autophagy; cysteine protease involved in autophagy APG4-D
Gene Symbols Atg4d
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATG4D(Accession Q86TL0), corresponding to amino acid residues Y425-L474.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235040, 84971
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less