missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATG4B Recombinant Rabbit Monoclonal Antibody (23GB3240)
GREENER_CHOICE

Catalog No. MA553612
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553612 100 μL
MA553613 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553612 Supplier Invitrogen™ Supplier No. MA553612
Only null left

Rabbit Recombinant Monoclonal Antibody

ATG4B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Cysteine protease ATG4b is an enzyme which is coded by ATG4b gene. Autophagy is a critical cellular homeostatic process that controls the turnover of damaged organelles and protein. ATG4bis a deubiquitin-like protease that plays dual roles in the core machinery of autophagy. ATG4b, cleaves the C-terminal amino acid of ATG8 family proteins MAP1LC3, GABARAPL1, GABARAPL2 and GABARAP, to reveal a C-terminal glycine. Exposure of the glycine at the C-terminus is essential for ATG8 proteins conjugation to phosphatidylethanolamine (PE) and insertion to membranes, which is necessary for autophagy. ATG4b is mainly expressed in the skeletal muscle, brain, heart, liver and pancreas.

Specifications

Antigen ATG4B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3240
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Atg4b
Gene Accession No. Q8BGE6, Q9Y4P1
Gene Alias 2510009N07Rik; APG4 (ATG4) autophagy-related homolog B; APG4 autophagy 4 homolog B; APG4B; ATG4 autophagy related 4 homolog B; Atg4b; atg4b protein; Atg4bl; AUT2B; Aut2B1; Aut2b2; AUTL1; AUT-like 1 cysteine; AUT-like 1 cysteine endopeptidase; AUT-like 1, cysteine endopeptidase; autophagin 1; autophagin-1; Autophagin-2B; autophagy related 4 homolog B; autophagy related 4B cysteine peptidase; autophagy related 4B, cysteine peptidase; autophagy-related 4B; autophagy-related cysteine endopeptidase 1; Autophagy-related cysteine endopeptidase 2B; autophagy-related protein 4 homolog B; AW048066; bAut2B; Cysteine protease ATG4B; cysteine protease involved in autophagy APG4-B; hAPG4B; KIAA0943
Gene Symbols Atg4b
Host Species Rabbit
Immunogen A synthetic peptide of human Atg4B (250-300AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23192, 316640, 66615
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less