missing translation for 'onlineSavingsMsg'
Learn More

Medisca Atenolol USP

Catalog No. 14185286
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185286 100 g
14186453 25 g
14185401 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14185286 Supplier Medisca Supplier No. 026205*

Atenolol USP is a white to practically white, odorless powder.

Atenolol USP is a white to practically white, odorless powder.

Specifications

Chemical Name or Material Atenolol USP
Packaging Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 29122-68-7
Type Drug Standard
Molecular Weight (g/mol) 266.3388
Quantity 100 g
Percent Purity 98 to 102 %
Grade USP