missing translation for 'onlineSavingsMsg'
Learn More

ATBTA-Eu3+ 90.0+%, TCI America™

Catalog No. A208310MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A208310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A208310MG Supplier TCI America Supplier No. A208310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Europium fluorescent labeling reagent suitable for time-resolved fluorometry.

Chemical Identifiers

CAS 601494-52-4
Molecular Formula C37H30EuN6NaO8
Molecular Weight (g/mol) 861.64
MDL Number MFCD10566875
InChI Key WPJHPWPASHGOEZ-UHFFFAOYSA-J
Synonym Sodium [4′C-(4′C-Amino-4-biphenylyl)-2,2′C:6′C,2′C′C-terpyridine-6,6′C′C-diylbis(methyliminodiacetato)]europate(III)
PubChem CID 44629911
IUPAC Name europium(3+) sodium 2-({[6-(4-{4'-amino-[1,1'-biphenyl]-4-yl}-6'-{[bis(carboxylatomethyl)amino]methyl}-[2,2'-bipyridin]-6-yl)pyridin-2-yl]methyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Eu+3].NC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC(=NC(=C1)C1=NC(CN(CC([O-])=O)CC([O-])=O)=CC=C1)C1=NC(CN(CC([O-])=O)CC([O-])=O)=CC=C1

Specifications

Quantity 10 mg
Formula Weight 861.63
Percent Purity ≥90.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material ATBTA-Eu3+