accessibility menu, dialog, popup

UserName

NNOOOONNNOOOOH2NNaEu

CAS RN 601494-52-4

IUPAC Name: europium(3+) sodium 2-({[6-(4-{4'-amino-[1,1'-biphenyl]-4-yl}-6'-{[bis(carboxylatomethyl)amino]methyl}-[2,2'-bipyridin]-6-yl)pyridin-2-yl]methyl}(carboxylatomethyl)amino)acetate
Synonyms: europium(3+) sodium 2-({[6-(4-{4'-amino-[1,1'-biphenyl]-4-yl}-6'-{[bis(carboxylatomethyl)amino]methyl}-[2,2'-bipyridin]-6-yl)pyridin-2-yl]methyl}(carboxylatomethyl)amino)acetate
Molecular Weight (g/mol): 861.64
Molecular Formula: C37H30EuN6NaO8
InChi Key: WPJHPWPASHGOEZ-UHFFFAOYSA-J
SMILES: [Na+].[Eu+3].NC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC(=NC(=C1)C1=NC(CN(CC([O-])=O)CC([O-])=O)=CC=C1)C1=NC(CN(CC([O-])=O)CC([O-])=O)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results