missing translation for 'onlineSavingsMsg'
Learn More

Astrazon Red 6B, TCI America™

Catalog No. a086825g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A086825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A086825G Supplier TCI America Supplier No. A086825G
Only null left

Chemical Identifiers

CAS 6441-82-3
Molecular Formula C24H30Cl2N2
Molecular Weight (g/mol) 417.42
MDL Number MFCD00059990
InChI Key JQZWHMOVSQRYRN-UHFFFAOYSA-M
Synonym Stenacrile Brilliant Red 6B, Basic Violet 7
PubChem CID 6538422
IUPAC Name 2-(2-{4-[(2-chloroethyl)(ethyl)amino]-2-methylphenyl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride
SMILES [Cl-].CCN(CCCl)C1=CC=C(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)C(C)=C1

Specifications

Color Blue-Purple
Quantity 25 g
Formula Weight 417.42
Physical Form Crystalline Powder
Chemical Name or Material Astrazon Red 6B
EINECSNumber (5)-1972
TSCA Yes