accessibility menu, dialog, popup

UserName

CAS RN 6441-82-3

ClH3CNClNH3CCH3CH3CH3(E/Z)

CAS RN 6441-82-3

IUPAC Name: 2-(2-{4-[(2-chloroethyl)(ethyl)amino]-2-methylphenyl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride
Synonyms: 2-(2-{4-[(2-chloroethyl)(ethyl)amino]-2-methylphenyl}ethenyl)-1,3,3-trimethylindol-1-ium chloride
Molecular Weight (g/mol): 417.42
Molecular Formula: C24H30Cl2N2
InChi Key: JQZWHMOVSQRYRN-UHFFFAOYSA-M
SMILES: [Cl-].CCN(CCCl)C1=CC=C(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)C(C)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results