missing translation for 'onlineSavingsMsg'
Learn More

Astrazon Brilliant Red 4G, TCI America™

Catalog No. A086525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A086525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A086525G Supplier TCI America Supplier No. A086525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12217-48-0
Molecular Formula C23H26ClN3
Molecular Weight (g/mol) 379.932
MDL Number MFCD00071429
InChI Key NJIRSTSECXKPCO-UHFFFAOYSA-M
Synonym Stenacrile Brilliant Red 4G, Basic Red 14
PubChem CID 6435842
IUPAC Name 3-[N-methyl-4-[(E)-2-(1,3,3-trimethylindol-1-ium-2-yl)ethenyl]anilino]propanenitrile;chloride
SMILES CC1(C2=CC=CC=C2[N+](=C1C=CC3=CC=C(C=C3)N(C)CCC#N)C)C.[Cl-]

Specifications

Color Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Astrazon Brilliant Red 4G