CAS RN 12217-48-0
IUPAC Name:
2-(2-{4-[(2-cyanoethyl)(methyl)amino]phenyl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride
Synonyms:
2-(2-{4-[(2-cyanoethyl)(methyl)amino]phenyl}ethenyl)-1,3,3-trimethylindol-1-ium chloride
Molecular Weight (g/mol):
379.93
Molecular Formula:
C23H26ClN3
InChi Key:
NJIRSTSECXKPCO-UHFFFAOYSA-M
SMILES:
[Cl-].CN(CCC#N)C1=CC=C(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)C=C1
1
–
1
of
1
results
1
Astrazon Brilliant Red 4G, TCI America™
CAS: 12217-48-0 Molecular Formula: C23H26ClN3 Molecular Weight (g/mol): 379.932 MDL Number: MFCD00071429 InChI Key: NJIRSTSECXKPCO-UHFFFAOYSA-M Synonym: Stenacrile Brilliant Red 4G, Basic Red 14 PubChem CID: 6435842 IUPAC Name: 3-[N-methyl-4-[(E)-2-(1,3,3-trimethylindol-1-ium-2-yl)ethenyl]anilino]propanenitrile;chloride SMILES: CC1(C2=CC=CC=C2[N+](=C1C=CC3=CC=C(C=C3)N(C)CCC#N)C)C.[Cl-]