Learn More
Troemner™ ASTM Class 2 Weight with ISO/IEC 17025 Accredited Certificate
Troemener Individual Calibration Weights are Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
$335.02 - $617.65
Specifications
| Product Type | Calibration Mass |
|---|---|
| Certifications/Compliance | ISO/IEC 17025 Accredited Certificate |
| Material | Troemner Alloy 8 |
| Case Material | Polycarbonate |
| Shape | Cylindrical |
Description
- Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
- Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
- Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
Specifications
| Calibration Mass | |
| Troemner Alloy 8 | |
| Cylindrical |
| ISO/IEC 17025 Accredited Certificate | |
| Polycarbonate |