Learn More
Cytiva Assembly
Catalog No. 361041163
Catalog No. 361041163 Supplier Cytiva Supplier No. 29024065
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cable and Bowl assemblies for the Frac30
Assemblies for the Frac30 fraction collector for use in the AKTA start automated protein purification system.
Specifications
For Use With (Application) Protein Purification
For Use With (Equipment) ÄKTA Start
Quantity 1 Each