Learn More
United Scientific™ Aspirator Bottles
Heavy duty for safe storage.
Supplier:  United Scientific™ 34101
 View more versions of this product
Catalog No. S15949
Only null left
Description
Description
  • These lightweight, autoclavable aspirator bottles are made from polypropylene and can be used for dispensing distilled water and other laboratory solutions.
  • The leak-proof spigot screwed to the bottle requires only a half turn to open or close.
  • Polyethylene cap and stopcock should be removed before autoclaving.
Specifications
Chemical Identifiers
White
Non-sterile
Aspirator Bottle
Polypropylene
Cylindrical
Autoclavable
Specifications
5 L
Polypropylene
Cylindrical
Autoclavable
White
Non-sterile
Aspirator Bottle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm