Learn More
United Scientific™ Aspirator Bottles
Heavy duty for safe storage.
Supplier: United Scientific™ 34102
Description
- These lightweight, autoclavable aspirator bottles are made from polypropylene and can be used for dispensing distilled water and other laboratory solutions.
- The leak-proof spigot screwed to the bottle requires only a half turn to open or close.
- Polyethylene cap and stopcock should be removed before autoclaving.
Chemical Identifiers
| White | |
| Non-sterile | |
| Aspirator Bottle |
| Polypropylene | |
| Cylindrical | |
| Autoclavable |
Specifications
| 10 L | |
| Polypropylene | |
| Cylindrical | |
| Autoclavable |
| White | |
| Non-sterile | |
| Aspirator Bottle |
Warning: Cancer and reproductive harm