missing translation for 'onlineSavingsMsg'
Learn More

Aspartame, Powder, FCC, 98-102%, Spectrum™ Chemical

A1377, 22839-47-0, C14H18N2O5

$247.15 - $1,756.60

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
IUPAC Name (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18600794
View Documents Promotion Details
N/A 25 g Poly Bottle
Each for $247.15
Only null left
 
18600795
View Documents Promotion Details
N/A 125 g Poly Bottle
Each for $648.58
Only null left
 
18600796
View Documents Promotion Details
N/A 500 g Poly Bottle
Each for $1,756.60
Only null left
 
Description

Description

Spectrum™ Chemical Aspartame, Powder, FCC is a well known sugar substitute and artificial sweetener that is widely used in food and drinks. It much sweeter than sucrose and leaves a longer lasting taste. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

22839-47-0
294.31
IAOZJIPTCAWIRG-QWRGUYRKSA-N
COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
C14H18N2O5
MFCD00002724
(3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid

Specifications

22839-47-0
0.2%
4.5%
MFCD00002724
COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
294.31
98 to 102%
100%
1 mg/kg
C14H18N2O5
IAOZJIPTCAWIRG-QWRGUYRKSA-N
(3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
+14.5° to +16.5°
FCC
Videos
SDS
Documents

Documents