missing translation for 'onlineSavingsMsg'
Learn More

Aspartame, Powder, FCC, 98-102%, Spectrum™ Chemical

Catalog No. 18600794
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600794 25 g Poly Bottle
18600795 125 g Poly Bottle
18600796 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600794 Supplier Spectrum Chemical Mfg Cor Supplier No. A137725GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1377, 22839-47-0, C14H18N2O5

Spectrum™ Chemical Aspartame, Powder, FCC is a well known sugar substitute and artificial sweetener that is widely used in food and drinks. It much sweeter than sucrose and leaves a longer lasting taste. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
IUPAC Name (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Lead (Pb) 1 mg/kg
Loss on Drying 4.5%
Packaging Poly Bottle
Quantity 25 g
Optical Rotation +14.5° to +16.5°
Percent Purity 98 to 102%
Grade FCC