missing translation for 'onlineSavingsMsg'
Learn More

Aspartame, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111013600
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013600 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013600 Supplier Merck Emd Millipore Supplier No. PHR13811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Aspartame is a low-calorie nutritive sugar substitute used in table-top sweeteners, chewing gums, soft drinks, etc.

Specifications

CAS 22839-47-0
Physical Form Neat
Linear Formula HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
MDL Number MFCD00002724
Synonym Asp-Phe methyl ester; N-(L-alpha-Aspartyl)-L-phenylalanine methyl ester; Asp-Phe-OMe; Aspartame
Quantity 1 g
Formula Weight 294.30
Grade Certified Reference Material