missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Asparagine Synthetase Recombinant Rabbit Monoclonal Antibody (23GB1475)

Catalog No. MA553171
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553171 20 μL
MA553170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553171 Supplier Invitrogen™ Supplier No. MA553171
Only null left

Rabbit Recombinant Monoclonal Antibody

Asparagine Synthetase Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

ASNS (asparagine synthetase [glutamine-hydrolyzing]) is involved in the subpathway that synthesizes f L-asparagine from L-aspartate. Mutations affecting the gene can result in asparagine synthetase deficiency (ASNSD).

Specifications

Antigen Asparagine Synthetase
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1475
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Asns
Gene Accession No. P08243, P49088, Q61024
Gene Alias ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein
Gene Symbols Asns
Host Species Rabbit
Immunogen A synthesized peptide derived from human ASNS (500-561AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25612, 27053, 440
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less