missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Asparagine Synthetase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150002
Description
Asparagine Synthetase Polyclonal Antibody for Western Blot, ICC/IF, IHC
ASNS (asparagine synthetase [glutamine-hydrolyzing]) is involved in the subpathway that synthesizes f L-asparagine from L-aspartate. Mutations affecting the gene can result in asparagine synthetase deficiency (ASNSD).Chemical Identifiers
| Asparagine Synthetase | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08243, P49088, Q61024 | |
| 25612, 27053, 440 | |
| Recombinant Asparagine Synthetase (ASNS) (His213-Tyr536). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Asns | |
| ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein | |
| Asns | |
| Primary | |
| Protein A |
Specifications
| Asparagine Synthetase | |
| Polyclonal | |
| Unconjugated | |
| Asns | |
| ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 25612, 27053, 440 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08243, P49088, Q61024 | |
| Asns | |
| Recombinant Asparagine Synthetase (ASNS) (His213-Tyr536). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |