missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASF1B Monoclonal Antibody (K.860.0)
Rabbit Monoclonal Antibody
Supplier: Invitrogen™ MA514836
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with ASF1A protein.
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The encoded protein is the substrate of the tousled-like kinase family of cell cycle-regulated kinases, and may play a key role in modulating the nucleosome structure of chromatin by ensuring a constant supply of histones at sites of nucleosome assembly.Specifications
| ASF1B | |
| Monoclonal | |
| 11 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q9NVP2 | |
| ASF1B | |
| Synthetic peptide corresponding the the carboxy-terminus of the human ASF1B protein. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| K.860.0 | |
| Unconjugated | |
| ASF1B | |
| 1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55723 | |
| -20°C | |
| Liquid |