missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Ascorbyl Palmitate NF
Description
Ascorbyl Palmitate NF is a white to yellowish white powder, having a characteristic odor.
Alternative name(s): Palmitoyl l-ascorbic
Specifications
Specifications
| Chemical Name or Material | Ascorbyl Palmitate NF |
| Packaging | Jar, 600mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 137-66-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 414.5352 |
| Quantity | 100 g |
| Percent Purity | 95 to 100.5 % |
| Grade | NF |