missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7157024
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600777 25 g Amber Glass Bottle
18600782 100 g Amber Glass Bottle
18600778 500 g Amber Glass Bottle
18600779 1 kg Amber Glass Bottle
18600780 5 kg Poly Pail
18600781 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600777 Supplier Spectrum Chemical Mfg Cor Supplier No. AS10225GM
Only null left

AS102, 50-81-7, C6H8O6

Spectrum™ Chemical Ascorbic Acid, USP , Vitamin C, is a dietary supplement used to treat and prevent vitamin C deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +20.5° to +21.5°
Percent Purity 99 to 100.5%
Grade USP