missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid, USP, 99-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Ascorbic Acid, USP , Vitamin C, is a dietary supplement used to treat and prevent vitamin C deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.1% |
| Packaging | Amber Glass Bottle |
| Quantity | 100 g |
| Specific Rotation | +20.5° to +21.5° |
| Percent Purity | 99 to 100.5% |
| Grade | USP |