missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7156939
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600773 100 g Amber Glass Bottle
18600774 500 g Amber Glass Bottle
18600776 5 kg Poly Pail
18600775 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600773 Supplier Spectrum Chemical Mfg Cor Supplier No. A1370100GM
Only null left

A1370, 50-81-7, C6H8O6

Spectrum™ Chemical Ascorbic Acid, Reagent, ACS , is an organic compound that occurs naturally and has antioxidant properties. It is a form of vitamin C and is classed as a reductone and used as an antioxidant food additive. Other uses include reactant in photographic developer solutions, antioxidant in fluorescence microscopy to increase fluorescent signals and chemically retard dye photobleaching. As an ACS grade Reagent, Spectrum Chemical manufactured Ascorbic Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +20.5° to +21.5°
Percent Purity 99%
Grade ACS