missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18600776
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
12 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600776 5 kg Amber Glass Bottle
18600773 100 g Amber Glass Bottle
18600774 500 g Amber Glass Bottle
18600775 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600776 Supplier Spectrum Chemical Mfg Cor Supplier No. A13705KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A1370, 50-81-7, C6H8O6

Spectrum™ Chemical Ascorbic Acid, Reagent, ACS , is an organic compound that occurs naturally and has antioxidant properties. It is a form of vitamin C and is classed as a reductone and used as an antioxidant food additive. Other uses include reactant in photographic developer solutions, antioxidant in fluorescence microscopy to increase fluorescent signals and chemically retard dye photobleaching. As an ACS grade Reagent, Spectrum Chemical manufactured Ascorbic Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 kg
Specific Rotation +20.5° to +21.5°
Percent Purity 99%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Ascorbic Acid