missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid (Powder/USP/FCC), Fisher Chemical™

Catalog No. p-61096
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A62500 500 g Poly Bottle
A62212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A62500 Supplier Thermo Fisher Scientific Supplier No. FLA62 500
Only null left

Not for human use

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 190°C
Color Yellow
Density 1.65g/cm³
Ignition Residue 0.1% max. (FCC, USP)
Packaging Poly Bottle
Linear Formula CH2OHCHOHCHCOH:COHCOO
Quantity 500 g
Specific Rotation +20.5 to +21.5° (FCC, USP)
Residual Solvents Meets Requirements (USP)
Formula Weight 176.13
Percent Purity 99.0 to 100.5% (USP)
Grade FCC/USP
Physical Form Solid
Chemical Name or Material Ascorbic Acid
Show More Show Less