missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, Fine Powder, 200 to 325 Mesh, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. p-5391065
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002364 500 g Wide Mouth Amber Glass Bottle
02002363 1 kg Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002364 Supplier Avantor Supplier No. 093907
Only null left

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Mesh Size 200 to 325 Mesh
Packaging Wide Mouth Amber Glass Bottle
Quantity 500 g
Specific Gravity 1L = 1.65kg
Formula Weight 176.12
Grade Multi-Compendial/USP
Physical Form Fine Powder
Chemical Name or Material Ascorbic Acid