missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid, Fine Granular, 20 to 80 Mesh, U.S.P. - F.C.C., J.T. Baker™
Supplier: Avantor 093808
Specifications
| Ascorbic Acid | |
| 20 to 80 Mesh | |
| C6H8O6 | |
| 1 kg | |
| l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
| OC[C@H](O)[C@H]1OC(=O)C(O)=C1O | |
| 176.12 | |
| CHEBI:29073 | |
| FCC/USP |
| 50-81-7 | |
| Poly Bottle | |
| MFCD00064328 | |
| 1L = 1.65kg | |
| CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one | |
| 54670067 | |
| 176.12 | |
| Fine Granular |
Chemical Identifiers
| 50-81-7 | |
| 176.12 | |
| CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
| 54670067 | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
| C6H8O6 | |
| MFCD00064328 | |
| l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
| CHEBI:29073 | |
| OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |