missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic acid, 99.0%, ACS Grade, MilliporeSigma™

Catalog No. MAX17753
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX17753 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX17753 Supplier Technidata Supplier No. AX17753

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Assay Percent Range 99.00%
Packaging Poly bottle
Linear Formula C6H8O6
Quantity 100 g
Formula Weight 176.13
Grade ACS
Chemical Name or Material Ascorbic acid
Recommended Storage Room Temperature