missing translation for 'onlineSavingsMsg'
Learn More

Ascomycin Solution, 1.0 mg/mL in Acetonitrile, Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. A0941ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0941ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0941ML Supplier MilliporeSigma Supplier No. A0941ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • An internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus.
  • This Certified Spiking Solution™ is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the narrow therapeutic range of these immunosuppressants.
  • The Snap-N-Spike™ format provides laboratories the ability to spike into their matrix of choice just before use and allows them to eliminate weighing operations of hazardous substances which significantly reduces occupational exposure hazards related to handling acutely toxic powders such as immunosuppressants.

Specifications

Chemical Name or Material Ascomycin
Packaging Ampule
CAS 104987-12-4
Molecular Formula C43H69NO12
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Clinical Testing
Molecular Weight (g/mol) 792.01
Quantity 1 mL
Grade Certified
Show More Show Less