missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144669 Supplier Invitrogen™ Supplier No. PA5144669
Only null left

Rabbit Polyclonal Antibody

Positive control: A549 cell lysate, Hela cell lysate, U87MG cell lysate, 293T cell lysate, HL-60 cell lysate, HepG2 cell lysate, rat liver tissue lysates. Predicted band size: 66 kDa Subcellular Location: Nucleus, Cytoplasm, cytosol, Cytoplasm, cytoskeleton, microtubule organizing center, centrosome.

Transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. Plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. Acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. Plays a role in androgen receptor transactivation and in testicular function.

Specifications

Antigen ASC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Trip4
Gene Accession No. Q15650
Gene Alias 4930558E03Rik; Activating signal cointegrator 1; Asc1; ASC-1; BB191711; HsT17391; SMABF1; thyroid hormone receptor interactor 4; thyroid receptor-interacting protein 4; transcription coactivator Asc1; TR-interacting protein 4; TRIP4; TRIP-4; ZC2HC5; zinc finger, C2HC5-type
Gene Symbols Trip4
Host Species Rabbit
Immunogen Recombinant protein within human ASC1 aa 1-581/581.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315769, 9325
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less