missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ASB-16 Detergent, Calbiochem™,

Catalog No. 18275525GM
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18275525GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18275525GM Supplier MilliporeSigma™ Supplier No. 18275525GM
Only null left

Chemical Identifiers

CAS 52562-29-5
Molecular Formula C24H50N2O4S
Molecular Weight (g/mol) 462.73
MDL Number MFCD03791122
InChI Key CLCSYZQBLQDRQU-UHFFFAOYSA-N
Synonym Amidosulfobetaine
PubChem CID 5067680
IUPAC Name 3-[(3-hexadecanamidopropyl)dimethylazaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCCCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Packaging Glass Bottle
Percent Purity ≥95% (TLC)