accessibility menu, dialog, popup

UserName

H3CONHNH3CCH3SOOO

CAS RN 52562-29-5

IUPAC Name: 3-[(3-hexadecanamidopropyl)dimethylazaniumyl]propane-1-sulfonate
Synonyms: 3-[(3-hexadecanamidopropyl)dimethylammonio]propane-1-sulfonate
Molecular Weight (g/mol): 462.73
Molecular Formula: C24H50N2O4S
InChi Key: CLCSYZQBLQDRQU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results