missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Specifications
| Antigen | ASAP1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine
ASAP1, also known as Development and differentiation enhancing factor 1, is a protein with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. ASAP1 is a homodimer known to interact with SRC and CRK and its activity is stimulated by phosphatidylinositol 4,5-biphosphate (PIP2). It possesses phosphatidylinositol 4,5-biphosphate-dependent GTPase-activating protein activity for ARF1 (ADP ribosylation factor 1) and ARF5 and a lesser activity towards ARF6. It may coordinate membrane trafficking with cell growth or actin cytoskeleton remodeling by binding to both SRC and PIP2 and may also function as a signal transduction protein involved in the differentiation of fibroblasts into adipocytes and possibly other cell types. It is widely expressed in most of the tissues. ASAP1 is a protooncogene involved in invasive breast cancer and in other types of invasive and maligt tumors, such as uveal melanomas.Specifications
| ASAP1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| AV239055, Ddef1, PAP, mKIAA1249, s19 | |
| ASAP1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9ULH1 | |
| 50807 | |
| Between 700 and 750 | |
| Primary | |
| 4° C | |
| ASAP1 |