missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557372 Supplier Invitrogen™ Supplier No. PA557372
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: PVDKYIEYDP VIILINAILC KAQAYRHILF NTQINIHGKL CIFCLLCEAY LRWWQLQDSN QNTAPDDLIR YAKEW Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

May act as a mediator of sterol homeostasis.

Specifications

Antigen ARV1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARV1
Gene Accession No. Q9H2C2
Gene Alias 1110067L22Rik; AI461928; Arv1; ARV1 fatty acid homeostatsis modulator; ARV1 homolog (S. cerevisiae); ARV1 homolog (yeast); ARV1 homolog, fatty acid homeostasis modulator; ARV1 homolog, fatty acid homeostatsis modulator; AW121084; hARV1; HT035; Protein ARV1; RGD1310264
Gene Symbols ARV1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARV1. Recombinant protein control fragment (Product #RP-103709).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less