missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Articaine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 23964-58-1 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C13H20N2O3 S |
| Synonym | Articaine; Carticaine; Methyl 4-methyl-3-[2-(propylamino)propionamido]-2-thiophenecarboxylate |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 284.37 |
| Grade | Certified Reference Material |