Learn More
Artesunate, Thermo Scientific Chemicals
$82.92 - $255.17
Chemical Identifiers
| CAS | 88495-63-0 |
|---|---|
| Molecular Formula | C19H28O8 |
| Molecular Weight (g/mol) | 384.42 |
| InChI Key | FIHJKUPKCHIPAT-RVBFJZMGSA-N |
| Synonym | artesunate |
| PubChem CID | 129317578 |
| SMILES | CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC(=O)CCC(=O)O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC460340050
|
Thermo Scientific Chemicals
460340050 |
5 g |
Each for $82.92
|
|
|||||
|
AC460340250
|
Thermo Scientific Chemicals
460340250 |
25 g |
Each for $255.17
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 88495-63-0 | |
| 384.42 | |
| artesunate | |
| CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC(=O)CCC(=O)O)C |
| C19H28O8 | |
| FIHJKUPKCHIPAT-RVBFJZMGSA-N | |
| 129317578 |
Specifications
| 88495-63-0 | |
| C19H28O8 | |
| artesunate | |
| CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC(=O)CCC(=O)O)C | |
| 129317578 | |
| Artesunate |
| 131.0°C to 135.0°C | |
| 5 g | |
| FIHJKUPKCHIPAT-RVBFJZMGSA-N | |
| 384.42 | |
| 384.42 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
GHS Signal Word: Warning
RUO – Research Use Only